Human recombinant Amiloride-binding 1 (from cells)
Leverandør: ProSci Inc.
92-393
PRSI92-393EA 5025 NOK
PRSI92-393
Human recombinant Amiloride-binding 1 (from cells)
Proteiner
Amiloride-sensitive amine oxidase (AOC1) belongs to the copper/topaquinone oxidase family. The protein exists as homodimer by disulfide and mainly located in placenta and kidney. AOC1 catalyses the degradation of compounds such as putrescine, histamine, spermine, and spermidine, substances involved in allergic and immune responses, cell proliferation, tissue differentiation, tumour formation, and possibly apoptosis. Placental DAO is thought to play a role in the regulation of the female reproductive function. The activity of this protein can be inhibited by amiloride in a competitive manner. It is inhibited by amiloride, a diuretic that acts by closing epithelial sodium ion channels.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.