Human recombinant FABP6 (from E. coli)
Leverandør: ProSci Inc.
96-294
PRSI96-294EA 7225 NOK
PRSI96-294
Human recombinant FABP6 (from E. coli)
Proteiner
Fatty acid-binding protein 6 (FABP6), is also known as Intestinal bile acid-binding protein (I-BABP), Ileal lipid-binding protein (ILBP), Intestinal 15 kDa protein (I-15P), Gastrotropin (GT). FABP6 is a cytoplasm protein which belongs to the calycin superfamily and fatty-acid binding protein (FABP) family. Isoform 2 of FABP6 is expressed in colorectal adenocarcinomas and their adjacent normal mucosa (at protein level). Isoform 1 of FABP6 is expressed in the jejunum, ileum, cecum and ascending colon intestine. Ileal protein FABP6 stimulates gastric acid and pepsinogen secretion. FABP6 seems to be able to bind to bile salts and bilirubins. Isoform 2 of FABP6 is essential for the survival of colon cancer cells to bile acid-induced apoptosis.