Anti-IL13 Rabbit Polyclonal Antibody

Leverandør: ProSci Inc.
XP-5168
PRSIXP-5168EA 6275 NOK
PRSIXP-5168
Anti-IL13 Rabbit Polyclonal Antibody
Antistoffer
L13 inhibits proinflammatory cytokine production and stimulates antibody production. It induces proliferation in the human pre myeloid cell line TF1. IL13 has multiple effects on the differentiation and functions of monocytes and macrophages. It suppresses cytotoxic functions and induces changes in the morphology of human monocytes and in the phenotype of human monocytes and B cells by upregulating MHC class II expression. IL13 will also decrease the production of nitric oxide by activated murine macrophages, leading to impaired parasiticidal activity. Human and mouse interleukin 13 shareMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol IL13
Environmentally Preferable
Antigen-navn Interleukin 13
Antigen synonymer MGC116786|ALRH|MGC116789|IL-13|BHR1|interleukin-13|MGC116788|P600
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Mouse
Vert Rabbit
Isotype
Western blot Yes
ELISA Yes
Nøytralisering Yes
Immunogen Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant mIL-13 (murine Interleukin-13).
Skjema Lyophilized

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR