Anti-IL2RB Rabbit Polyclonal Antibody (Biotin)
Leverandør: ProSci Inc.
XP-5169BT
PRSIXP-5169BTEA 6275 NOK
PRSIXP-5169BT
Anti-IL2RB Rabbit Polyclonal Antibody (Biotin)
Antistoffer
IL-15 (14-15 kD) is a member of the four alpha-helical bundle family of cytokines. It is very similar to IL-2, except that IL-15 has an IL-15 alpha receptor subunit. IL-15 plays an important role in the growth and differentiation of T and B lymphocytes, natural killer cells, macrophages, and monocytes as well as activation of a number of important intracellular signaling molecules. This implies that IL-15 could be essential for the immune responses, allograft rejection, and the pathogenesis of autoimmune diseases.
Anti-hIL-15 specific antibody was purified by affinity chromatography andMore Product Information