Anti-IL1A Rabbit Polyclonal Antibody

Leverandør: ProSci Inc.
XP-5174
PRSIXP-5174EA 6275 NOK
PRSIXP-5174
Anti-IL1A Rabbit Polyclonal Antibody
Antistoffer
Anti-hIL-1alpha specific antibody was purified by affinity chromatography employing immobilized hIL-1alpha matrix.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: IL1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR