Anti-AFP Chicken Polyclonal Antibody
Leverandør: ProSci Inc.
XW-7017
PRSIXW-7017EA 10575 NOK
PRSIXW-7017
Anti-AFP Chicken Polyclonal Antibody
Antistoffer
beta-1-fetoprotein. beta-fetoprotein is a major plasma protein produced by the yolk sac and the liver during fetal life. beta-fetoprotein expression in adults is often associated with hepatoma or teratoma. However, hereditary persistance of beta-fetoprotein may also be found in individuals with no obvious pathology. The protein is thought to be the fetal counterpart of serum albumin, and the beta-fetoprotein and albumin genes are present in tandem in the same transcriptional orientation on chromosome 4. beta-fetoprotein is found in monomeric as well as dimeric and trimeric forms, and bindsMore Product Information