Anti-HAGH Chicken Polyclonal Antibody

Leverandør: ProSci Inc.
XW-7892
PRSIXW-7892EA 10150 NOK
PRSIXW-7892
Anti-HAGH Chicken Polyclonal Antibody
Antistoffer
FUNCTION: Thiolesterase that catalyzes the hydrolysis of S-D-lactoyl-glutathione to form glutathione and D-lactic acid.
CATALYTIC ACTIVITY: S-(2-hydroxyacyl)glutathione + H2O = glutathione + a 2-hydroxy carboxylate.
SUBUNIT: Monomer.
SIMILARITY: Belongs to the glyoxalase II family.
COFACTOR: Binds 2 zinc ions per subunit.
PATHWAY: Glyoxal pathway.

Hydroxyacylglutathione hydrolase antibody can be used for the detection of Hydroxyacylglutathione hydrolase by Western Blot.

Phosphate-Buffered Saline. No preservatives added.

Phosphate-Buffered Saline. No preservatives added.

Aliquot andMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol HAGH
Environmentally Preferable
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human,Rat,Mouse
Vert Chicken
Isotype
Western blot Yes
Immunogen 110-260
Skjema Liquid
Konsentrasjon 1 mg/mL
Uttestet applikasjon Immunoaffinity Purified

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR