Human Leukopak
Leverandør: PROVITRO
12192100025 12192100050 12192100100 12192100200 11192100025 11192100050 11192100100 11192100200 12192200999 11192200999
PRTV12192100025EA 40175 NOK
PRTV12192100025 PRTV12192100050 PRTV12192100100 PRTV12192100200 PRTV11192100025 PRTV11192100050 PRTV11192100100 PRTV11192100200 PRTV12192200999 PRTV11192200999
Human Leukopak
Celler
Our highly cell-concentrated Leukopak is a specialised product from leukapheresis using the Spectra Optia system. This advanced technology employs centrifugal mechanisms for continuous blood flow, extracting white blood cells from peripheral blood into a sterile bag with ACD-A anticoagulant. The enriched product includes monocytes, lymphocytes, platelets, plasma, and erythrocytes.
  • Leukopak is collected from healthy donors using approved permission forms and procedures
  • Collection procedures have been approved by the Institutional Review Board (IRB)
  • The product includes donor information and serology testing for: HIV, HBs-AG, HCV
  • Additional optional information and testing are available for a small fee
Our offerings encompass Leukopaks sourced from our existing inventory, alongside the provision for prospective collections tailored to specific research inquiries. This versatile range ensures a breadth of options to suit a variety of research requirements, whether drawing from our readily available stock or arranging for custom collections based on unique project needs.
Storage: Prepare cells for extended storage in the vapor phase of liquid nitrogen or thaw them for immediate use. Avoid storing cells in the liquid phase of nitrogen. Short-term storage (less than two weeks) at −80 °C is acceptable but should be minimised for optimal stability. Once thawed, it's crucial to use the samples immediately.
NB: Not intended for any human or animal diagnostic or therapeutic use.