Skeletal Muscle Cell Growth Medium
Leverandør: PROVITRO
2110602
PRTV2110602EA 2325 NOK
PRTV2110602
Skeletal Muscle Cell Growth Medium
Media, Cellekultur
Skeletal Muscle Cell Growth Medium is a sterile liquid culture medium designed for cultivating human skeletal muscle cells (HSKMC). Delivered as a basal medium, it is suitable for culturing Provitro HSKMC upon the addition of supplement mix components.
  • Formulation is optimised for initial seeding of 4000 cells/cm², facilitating growth up to confluence (approximately 90%)
  • Medium does not require feeder layers, matrix substrates, or additional substances for effective cell cultivation
Due to the possibility of reduced proliferative activity, we recommend to use the antibiotic supplement for freshly isolated cells only. Skeletal muscle cell growth medium is thoroughly tested after each production. All components are tested in a stringent biological assay.
Each batch is checked for human endothelial cells proliferating characteristics. The cells cultured in endothelial cell growth medium are checked regarding their morphology, the adherence rate, the colony forming efficiency and the population doubling time.
Storge: Store the supplemented smooth muscle cells at a temperature of −20 °C. The supplements must not be used after the expiry date. Always adhere to these storage guidelines to maintain the effectiveness and integrity.