Cancer Stem Cell Medium (BIT-100)
Leverandør: PROVITRO
2131001
PRTV2131001EA 5125 NOK
PRTV2131001
Cancer Stem Cell Medium (BIT-100)
Media, Cellekultur
Cancer Stem Cell Medium is a sterile liquid culture medium designed for the cultivation of human cancer stem cells. Delivered in the form of a basal medium, it becomes a complete and effective environment for the growth of human cancer stem cells upon the addition of supplement mix components.
  • Formulation is fine-tuned for optimal performance, supporting an initial seeding of 6000 cells/cm² up to confluence (approx. 90%)
  • Feeder-layer, matrix substrates, or other substances are not necessary
  • Also used in cell culture, particularly in systems with a low serum content or completely serum-free culture systems
  • This product is derived from E.coli using human donors as the source
Due to the possibility of reduced proliferative activity, we recommend to use any antibiotic supplement for freshly isolated cells only. All components are tested in a stringent biological assay. Each batch is checked for human cancer stem cell proliferating characteristics. The cells cultured in cancer stem cell medium are checked regarding their morphology, the adherence rate, the colony forming efficiency, and the population doubling time.
Storge: Store the supplemented Cancer stemm cell medium in a dark environment at temperatures ranging from 4 to 8 °C for a maximum duration of one month. Avoid heating the medium beyond 37 °C and refrain from using uncontrolled heat sources, such as microwave appliances. When utilising a portion of the medium, extract the required amount from the bottle and warm it appropriately. Never freeze the medium, as this could result in elevated salt concentrations due to the separation of pure water during freezing, leading to irreversible damage.