Ear plugs, PU foam, detectable, EP30
Leverandør: Portwest
EP30BLU
PRTWEP30BLUEA 1700 NOK
PRTWEP30BLU
Ear plugs, PU foam, detectable, EP30
Ørepropper Ørepropper til engangsbruk
These detectable disposable ear plugs are comfortable to fit due to the soft PU foam material. Corded for increased safety and visibility in food industry environments. Each pair is inside a resealable hygienic polybag.
  • Can be used in food industries (metal detectable)
  • Retail box which aids presentation for retail sales
  • Individually packed for vending machines
These avoids the hearing problems occurring when working under hazardous conditions.
Sertifiseringer: EN 352-2 (SNR 37dB); ANSI S3.19 (32dB).
Forpakning: 200/pack, 10/carton.