Fall protection harnesses, 2 point comfort
Leverandør: Portwest
FP14RER
PRTWFP14REREA 875 NOK
PRTWFP14RER
Fall protection harnesses, 2 point comfort
Fallvern Fallsele
Fall Protection is put in place to prevent the risks associated with falling from heights, reducing impact force, restricting obstacle/ground collision and restricting users from fall hazard areas.
  • Harness with a sliding dorsal D-Ring
  • Rear D-ring for keys or ID cards
  • Chest adjustment point
  • Padded belt
  • Made from polyester webbing, alloy steel, heat treated, polyester mesh
Features a breathable contoured backpad, two side D-Rings and flat bar buckles which are easy to use and fail safe. The waist, chest and leg adjusters offer excellent fit and userMore Product Information