Goggles, indirect vent
Leverandør: Portwest
PW21CLR
PRTWPW21CLREA 67 NOK
PRTWPW21CLR
Goggles, indirect vent
Vernebriller /Goggles
Designed to be worn in a tough work environment, flexible PVC frame for ultra comfort, transparent colour for better side visibility.
  • 99% UV protection
  • Back elastic for ease of movement
  • Ultra lightweight for long uses
  • Individually packed for vending machines
  • Indirect ventilation
Sertifiseringer: EN 166 (1 B 3); ANSI/ISEA Z87.1