Endurance visor replacement, PW56
Leverandør: Portwest
PW56CLR
PRTWPW56CLREA 57 NOK
PRTWPW56CLR
Endurance visor replacement, PW56
Ansiktsskjermer
Impact resistant polycarbonate replacement visor is suitable for endurance helmets.
  • Anti-scratch coating for added durability
  • Anti-fog coating for additional comfort to the wearer
Suitable for PW54, PW55 and PS55 helmets.
Sertifiseringer: Standard: EN 166 1 FT.
Forpakning: Individually packed for vending machines.