Browguard with visor, PP
Leverandør: Portwest
PW91CLR
PRTWPW91CLREA 400 NOK
PRTWPW91CLR
Browguard with visor, PP
Ansiktsskjermer
PC visor with PP browguard, is manufactured from impact resistant PP.
  • Wheel ratchet adjustment system is designed for easy use
  • Replacement visor available (PW92)
Sertifiseringer: EN 166 (1 B/3 B)