Adapter kit
Leverandør: Cytiva
PSA410
PSA410EA 500 NOK
PSA410
Adapter kit
Strømkilder
Connects 4 mm output jack to 4 mm high voltage leads.
Note that this adaptor kit is not CE compliant.
Bestillingsinformasjon: Supplied with Allen wrench.