Chromium VI -WP, certified reference material, Supelco®
Leverandør: Merck
QC1088-20ML
QC1088-20MLEA 770 NOK
QC1088-20ML
Chromium VI -WP, certified reference material, Supelco®
Standarder Kationstandarder Chromium Standards
ICP and AAS suitable.
Values of analytes vary lot to lot.
Sertifiseringer: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.