Color-WP, certified reference material (CRM), Supelco®
Leverandør: Merck
QC1126-20ML
QC1126-20MLEA 835 NOK
QC1126-20ML
Color-WP, certified reference material (CRM), Supelco®
Standarder Fargereferanse og spekroskopistandarer
Sertifiseringer: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.