Trace Metals 1-WP, certified reference material, Supelco®
Leverandør: Merck
QC1132-20ML
QC1132-20MLEA 745 NOK
QC1132-20ML
Trace Metals 1-WP, certified reference material, Supelco®
Standarder Kationstandarder Multi-Element Standards
Analytes:
Aluminium; Arsenic; Beryllium;
Cadmium; Chromium; Cobalt; Copper;
Iron; Lead; Lithium;
Manganese; Mercury; Nickel;
Selenium; Vanadium; Zinc
Sertifiseringer: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.