Anti-Gentamicin Mouse monoclonal antibody unconjugated

Leverandør: QED Bioscience
16101
QEDB16101EA 4750 NOK
QEDB16101
Anti-Gentamicin Mouse monoclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: Gentamicin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR