Genomic DNA extraction from dried blood spots, Extracta™ DBS

Leverandør: Quantabio

95171-010 95171-500
QUNT95171-010EA 960 NOK
QUNT95171-010 QUNT95171-500
Genomic DNA extraction from dried blood spots, Extracta™ DBS
Nukleinsyre reagenser Nukleinsyrerensingsreagenser og -kit

Extracta™ DBS is a ready to use DNA extraction reagent for rapid and efficient recovery of PCR-ready DNA from dried blood spots (DBS) on Guthrie cards or Whatman® 903 filter/TFN or 226 from Ahlstrom Munksjö paper. This patented single-solution process produces DNA eluates that are substantially free of PCR inhibitors and compatible with a variety of end-point PCR, real-time PCR and Next Generation Sequencing (NGS) or Sanger Sequencing reagents. Application of Extracta DBS with PerfeCTa® qPCR ToughMix® or PerfeCTa MultiPlex qPCR ToughMix® enables accurate and reproducible quantification of DNA sequences in blood using TaqMan® hydrolysis probe real-time qPCR.


  • Optimised for DNA extraction from dried blood spot punches
  • Single reagent for PCR-ready DNA in 30 minutes
  • Maximised assay sensitivity, lower Cq values, when combined with Quantabio ToughMix®
  • Compatible with high throughput automation for PCR, qPCR and NGS applications

Bestill nå

SPESIFIKASJONER

Beskrivelse Extracta™ DBS
Environmentally Preferable
Applikasjon
Target molecule DNA
Refine target molecule Total DNA
Sample type Blood (dried) on filter cards
Method / Format Extraction for direct PCR
Prep size Scalable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR