Buffer, EDTA buffer solution pH 8,5 (0,5 mol/L)
Leverandør: G Bioscienes
R013
R013EA 1075 NOK
R013
Buffer, EDTA buffer solution pH 8,5 (0,5 mol/L)
Buffere Buffere, kloning
A DNase and RNase free solution of 0.5M EDTA.