Litium acetat 1 mol/l in aqueous solution

Leverandør: G Bioscienes

Synonymer: Acetic acid lithium salt, Lithiumacetate

R039
R039EA 1150 NOK
R039
Litium acetat 1 mol/l in aqueous solution
Litium acetat
Formel: H₃CCOOLi
MW: 65.99 g/mol
Smeltepunkt: 291
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00013057
CAS-nummer: 546-89-4
EINECS: 208-914-3
Merck Indeks: 12,05544

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR