RNA loading dye (2X)
Leverandør: Thermo Fisher Scientific
R0641
R0641EA 460 NOK
R0641
RNA loading dye (2X)
Farger Elektroforese Farging
In most denaturing agarose gel systems, bromophenol blue migrates slightly faster than human 5S rRNA, whereas xylene cyanol FF migrates slightly slower than 18S rRNA. RNA loading dye contains the denaturing agent formamide, which allows RNA fragments to separate according to size even during non-denaturing electrophoresis. Formamide also stabilises RNA.
  • Two-colour tracking of DNA, RNA migration during electrophoresis
  • No DNA, RNA masking during gel exposure to UV light
  • Denaturation of DNA, RNA due to the presence of formamide
Recommended for preparation of RNA samples for electrophoresis on agarose or polyacrylamide gels.
Bestillingsinformasjon: Contains the tracking dyes bromophenol blue and xylene cyanol FF, as well as the intercalating dye ethidium bromide.