Rhodamine 6G ~95% (dye content), Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Danger

Synonymer: 9-(2-(Ethoxycarbonyl)phenyl)-3,6-bis(ethylamino)-2,7-dimethylxanthylium chloride, 9-[2-(Ethoxycarbonyl)phenyl]-3,6-bis(ethylamino)-2,7-dimethylxanthylium chloride

M-Clarity™ quality level = MQ200

R4127-100G R4127-25G R4127-5G
R4127-100GEA 1150 NOK
R4127-100G R4127-25G R4127-5G
Rhodamine 6G ~95% (dye content), Sigma-Aldrich®
Rhodamine 6G

Rhodamine 6G has been used as a leukocyte marker to study the interaction between leukocytes and the vascular endothelium. It has also been used to label platelets.


Biochem/physiol Actions
Rhodamine 6G is a rhodamine analog useful in P-glycoprotein (Pgp) efflux assays. It has been used to characterize kinetics of multidrug resistance protein 1 (MRP1)-mediated efflux.
A rhodamine analog useful in Pgp efflux assays. Has been used to characterize kinetics of MRP1- mediated efflux.

Formel: C₂₈H₃₁ClN₂O₃
Kokepunkt: ? 200 °C (1013 hPa)
Smeltepunkt: 215 °C
Tetthet: 1,3 g/cm³
Lagringstemperatur: Room Temperature
MDL nummer: MFCD00012665
CAS-nummer: 989-38-8
EINECS: 213-584-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eea76600c9b7f6588fc55f5ec0813a33430725ae&vid=8623beeee65590d5599e861468de3859ad42bc0f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Solubility (colour) 1 mg/ml in water
Solubility (method) 1 mg/ml in water
Carbon content 66.5 - 74.0 %
Nitrogen content 5.0 - 6.2 %
Dye content ~ 95 %
Wavelength (1) (UV) ~ 525 nm
Extinction (1%/1 cm) (1) (UV) ≥ 1700
Solvent (UV) Water

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR