Rhodamin B ≥95% (by HPLC), Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Danger

Synonymer: 9-(2-Carboxyphenyl)-3,6-bis(diethylamino)xanthylium chloride, [9-(2-Carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium chloride

M-Clarity™ quality level = MQ200

R6626-100G R6626-25G R6626-500G R6626-100G R6626-25G R6626-500G
R6626-100GEA 906 NOK
R6626-100G R6626-25G R6626-500G SIALR6626-100G SIALR6626-25G SIALR6626-500G
Rhodamin B ≥95% (by HPLC), Sigma-Aldrich®
Rhodamin B

Rhodamine B is a xanthene dye, which functions as a water tracer fluorescent. It is used as a colorant in textiles and food stuffs. Rhodamine B is associated with eye, skin and respiratory tract irritation. It is used as a biomarker in oral rabies vaccine.


  • Rhodamine B has been used:
  • to represent fluorescence of tetra methyl rhodamine (TMR)
  • in the preparation of adhesive resin for confocal laser scanning microscopy
  • as a fluorescent dye in intrahemolymph dye dosing and retinal image acquisition
  • to depict brain-specific dye capture
  • for imaging targets

Formel: C₂₈H₃₁ClN₂O₃
Tetthet: 0,79 g/cm³
Lagringstemperatur: Room Temperature
MDL nummer: MFCD00011931
CAS-nummer: 81-88-9
EINECS: 201-383-9
Merck Indeks: 13,08266
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b559375693664cc78ffa204575387a72025cf342&vid=58d0fca01331f1b1a615ec381c631dac183e5206&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance (Colour) Faint Green to Very Dark Green
Appearance (Form) Powder
Solubility (Colour) Faint Red to Red and Faint Purple to Purple
Solubility (Turbidity), 1 mg/ml, H₂O Clear
UV Absorbance Ratio, ABS at 510 nm (UVAB)/ Lambda max (UVVS) ≤0.50
EmM 104.5 - 119.5
Lambda Max 542 - 554 nm
Purity (HPLC) ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR