Remazol brilliant blue R , Anthraquinone dye

Leverandør: SIGMA ALDRICH MICROSCOPY

Synonymer: Remazol Brilliant Blue R (C.I. 61200), Sodium 2-((3-((4-amino-9,10-dioxo-3-sulphonato-9,10-dihydroanthracen-1-yl)amino)phenyl)sulphonyl)ethyl sulphate, MFCD00001215, Reactive Blue 19, REACTIVEBLUE19

M-Clarity™ quality level = MQ200

R8001-25G
R8001-25GEA 645 NOK
R8001-25G
Remazol brilliant blue R , Anthraquinone dye
Remazol brilliant blue R

Remazol Brilliant Blue R (Cl 61200) is a hydrophobic, anthraquinone dye that has several commercial uses. It is a reactive dye that stains collagen, elastin, basement membranes, and erythrocytes. Remazol Brilliant Blue R is a hydrophobic, anthraquinone dye which has several commercial uses.


Remazol Brilliant Blue R has been used to prepare Remazol Brilliant blue (RBB)-starch agar medium to select and isolate amylolytic fungi. It has also been used to stain purified peptidoglycan for dye release assay.

Formel: C₂₂H₁₆N₂Na₂O₁₁S₃
MW: 626.533 g/mol
Lagringstemperatur: Room Temperature
MDL nummer: MFCD00001215
CAS-nummer: 2580-78-1

Bestill nå

Spesifikasjon Testresultater

Appearance (Form) Powder
Nitrogen 1.8 - 2.9 %
Conforms
40-65%; Calculated from Nitrogen
Dye Content
Solubility (Colour)
c = 1mg/ml; Water
Dark Blue
Recommended Retest Period 4 Years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR