Rose bengal diacetate, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY

R8401-5MG
R8401-5MGEA 2300 NOK
R8401-5MG
Rose bengal diacetate, Sigma-Aldrich®
Rose bengal diacetate

A cell-permeable generator of singlet oxygen.

Formel: C₂₄H₈Cl₄I₄O₇
Lagringstemperatur: Fryseskap
CAS-nummer: 61738-01-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab8ec578b70d3b568dc9138560283e08f5bc9a9b&vid=251ed10a1d03c6a9f5f6952a53eedf127d0871d7&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR