MCDh 4

Leverandør: RAL

313600-2500
RALA313600-2500EA 600 NOK
RALA313600-2500
MCDh 4
MCDh

MCDh 4 - RINSING SOLUTION Water based - 6% ethanol


  • MCDh (Micro Chromatic Detection for Hematology)
  • for RAL Stainer Kit (Ref 360200)
  • Methanol-free


Role
- Removes excess stain from cells (better differentiation)
- Finalizes the staining


MCDh is a panoptic May-Grünwald Giemsa (MGG) type staining method. Methanol free, it is made of a sequence of four specific reagents use in blood and bone marrow cell differentiation and counting. This new generation of stains is in line with the standardisation trend in diagnostic techniques. Unlike more traditional stains, all MCDh reagents are fast and ready to use. As no dilution is needed, consistency and standardisation are guaranteed.


Components - Name (ref) Packing
MCDH 1 (Ref 313590) 1L - 2,5L
MCDH 2 (Ref 313570) 1L - 2,5L
MCDH 3 (Ref 313560) 1L - 2,5L
MCDH 4 (Ref 313600) 1L - 2,5L

IVDLogo

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR