Gram-Nicolle Kit To differentiate Gram-positive from Gram-negative Bacteria

Leverandør: RAL

364320-0000
RALA364320-0000EA 1150 NOK
RALA364320-0000
Gram-Nicolle Kit To differentiate Gram-positive from Gram-negative Bacteria
Gram-Nicolle Kit

Gram-Nicolle staining is a differential staining based on the structure of the bacteria wall which is different whether it concerns Gram-positive bacteria or Gram-negative bacteria


Components - Name (ref) Packing
CARBOLIC GENTIAN VIOLET (Ref 364210) 240 ml (Other packagings 1000, 2500 ml)
LUGOL PVP STABILIZED SOLUTION (Ref 367400) 240 ml (Other packagings 1000, 2500 ml)
DIFFÉRENCIATOR FAST (Ref 361510) 240 ml (Other packagings 1000, 2500 ml)
ZIEHL FUCHSIN (Ref 364540) 240 ml (Other packagings 1000 ml, 2500 ml)

IVDLogo

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR