Workstation racks, PP
Leverandør: RATIOLAB
5300100
RATI5300100EA 830 NOK
RATI5300100
Workstation racks, PP
Stativer for rør Kryorørstativer
The cryotubes with self standing base are automatically locked in the workstation and can easily be screwed and unscrewed with one hand.
Made of high quality polypropylene with non-slip feet to stand safely on laboratory benches.