Cleanibag, ratiolab®
Leverandør: RATIOLAB
7000260 7000270 7000250
RATI7000260EA 2550 NOK
RATI7000260 RATI7000270 RATI7000250
Cleanibag, ratiolab®
Poser Prøveposer
All-purpose PE bags with self-adhesive strip for clinical and laboratory use.
  • Self-adhesive strip: sticks on smooth and clean surfaces (e.g. walls, work benches, beds, bedside tables)
  • Residue-free, removable
Bestillingsinformasjon: Other sizes or models available on request, e.g. for intensive care and bags with custom-made imprint.
Forpakning: 1000 pieces per cardboard box.