EDTA dinatrium salt dihydrat

Leverandør: G Bioscienes

RC-048
RC-048EA 5800 NOK
RC-048
EDTA dinatrium salt dihydrat
EDTA dinatrium salt dihydrat
Formel: C₁₀H₁₄N₂Na₂O₈·2H₂O
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00150037
CAS-nummer: 6381-92-6
EINECS: 205-358-3
Merck Indeks: 12,03556

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR