Refractix™ Universal Digital Refractometer
Leverandør: REICHERT
13910000
REIC13910000EA 33600 NOK
REIC13910000
Refractix™ Universal Digital Refractometer
Refraktometere Refraktometere, håndholdte
Refractometer provides laboratory performance in a compact portable platform.
  • Sapphire prism
  • 250 measurement scale capacity with 50 scales pre-loaded
  • 2,8" color touch screen and physical buttons
  • High resolution 1500 CCD linear image sensor with benchtop accuracy
  • Measurement log stores last 5000 measurements
  • Bluetooth and USB-C connectivity to RefractixSync PC software
  • Rechargeable battery with AC power option
  • Designed, engineered, and manufactured in the USA
The Refractix™ refractometer is innovative, thoughtfully designed, portable, and defiesMore Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Skalaer 250 scales
Nøyaktighet 0,0001 nD (0,1/100 g brix) [precision]
Måleområde 1,3300 - 1,5600 nD (0,0 - 85,0/100 g Brix ICUMSA)
Temperaturkompenseringområde 10...40 °C (50...104 °F)
Strømforsyning Rechargeable Li-ion battery and AC outlet power option
B×D×H 17,6×7,3×4,2 cm