Refractix™ Universal Digital Refractometer
Leverandør: REICHERT
13910000
REIC13910000EA 33600 NOK
REIC13910000
Refractix™ Universal Digital Refractometer
Refraktometere Refraktometere, håndholdte
Refractometer provides laboratory performance in a compact portable platform.
  • Sapphire prism
  • 250 measurement scale capacity with 50 scales pre-loaded
  • 2,8" color touch screen and physical buttons
  • High resolution 1500 CCD linear image sensor with benchtop accuracy
  • Measurement log stores last 5000 measurements
  • Bluetooth and USB-C connectivity to RefractixSync PC software
  • Rechargeable battery with AC power option
  • Designed, engineered, and manufactured in the USA
The Refractix™ refractometer is innovative, thoughtfully designed, portable, and defiesMore Product Information