Micropacked inlet conversion kits
Leverandør: Restek
22426
REST22426EA 2375 NOK
REST22426
Micropacked inlet conversion kits
Kromatografiutstyr Kromatografireservedeler
Micropacked inlet conversion kits are used to convert a capillary GC split/splitless inlet for use with ¹/₁₆" Ext.Ø micropacked columns.
  • For use with Agilent 5890 6890 and 7890 GCs
  • Sample pathways deactivated for ultimate inertness
Leveringsinformasjon: Kit includes dual large bore Vespel ring inlet seal, large bore reducing nut, large bore FID adaptor, ¹/₁₆" Vespel/graphite ferrule, ¹/₁₆" stainless steel nut, 4 mm splitless liner (intermediate polarity deactivated).