Flexible inspection light
Leverandør: Restek
22627
REST22627EA 1300 NOK
REST22627
Flexible inspection light
Lys LED-belysnings
This flexible inspeciton light inspects inside surfaces of sample cylinders or other chambers.
  • Has a reach of 355 mm
  • LED life of 100000 hours