Vial racks for headspace vials
Leverandør: Restek
22643
REST22643EA 785 NOK
REST22643
Vial racks for headspace vials
Stativer for rør Kromatografirørstativer
These vial racks securely hold up to 36 headspace vials.
  • Autoclavable up to 121 °C for 20 minutes
  • Vials lock in rack for convenient one-handed cap removal
  • Easy to clean
  • Stackable even with vials in place
The openings in the bottom of each well facilitate drainage and the alphanumeric indexing on rack simplifies sample identification.