Capillary dedicated FID replacement jet
Leverandør: Restek
21620 21621 23078
REST21620EA 1550 NOK
REST21620 REST21621 REST23078
Capillary dedicated FID replacement jet
Kromatografiutstyr Kromatografireservedeler
These capillary dedicated FID replacement jet for Agilent 5890/6850/6890/7890 GCs are available in standard, high-temperature or Siltek treated high-performance versions.
  • For standard version threads are specially coated for easy installation and removal
  • Special processing ensures the highest degree of cleanliness
  • High-performance version - Siltek treated are extremely inert for use with active compounds