Pack in a box kit
Leverandør: Restek
26408
REST26408EA 224100 NOK
REST26408
Pack in a box kit
Kromatografiutstyr Kromatografireservedeler
A complete column packing system which is used to pack the analytical HPLC columns.
  • Maximum pressure: 689 bar
  • AC input: 85 to 265 VAC
The packing pump is electric, so no pressurized air is required.
Leveringsinformasjon: This kit includes pump, 20 ml reservoir, column hardware (150×4,6 mm, 316 stainless steel tubing with end-fittings, frits, nuts and ferrules for two complete, empty columns), and system control software (computer not included). Detailed packing instructions and a DVD are also included.