Vibratory sieve shaker, AS 450 control
Leverandør: Retsch GmbH
30.026.0001
RETS30.026.0001EA 0 NOK
RETS30.026.0001
Vibratory sieve shaker, AS 450 control
Risteapparater Siktemaskiner
The analytical sieve shaker AS 450 control is used in research and development, quality control of raw materials, interim and finished products as well as in production monitoring. The controllable electromagnetic drive offers an optimal adaption for every product. Sharp fractions are obtainable even after very short
sieving times. It is suitable for dry and wet sieving.
  • Easy operation, ergonomic design and maintenance-free
  • Excellent separation efficiency even with short sieving times
  • CET Technology for controlled amplitude up to 2,2 mm even with high loads