Fluid bed dryer, TG 200
Leverandør: Retsch GmbH
70.760.0001
RETS70.760.0001EA 0 NOK
RETS70.760.0001
Fluid bed dryer, TG 200
Eksikatorer Sengetørker for væsker
The TG 200 fluid bed dryer is used in quality control, sample preparation and research departments. It allows the gentle drying of organic, inorganic, chemical or pharmaceutical bulk materials without localised overheating.
  • Gentle drying, dispersing and mixing also of temperature sensitive materials
  • Very short drying times (~ 5 to 20 min)
  • Powerful fan for optimal air throughput
  • Easy handling
  • Interval operation for better mixing of the fluidised bed
  • Wide range of accessories including various containers
  • Memory for up to 9 SOPs
  • Brushless motor provides long