Fiberboard cryobox dividers, Revco
Leverandør: REVCO TECHNOLOGIES
5958 5959 5960
REVC5958EA 455 NOK
REVC5958 REVC5959 REVC5960
Fiberboard cryobox dividers, Revco
Bokser Kryobokser
Fiberboard cryobox dividers for ultra-low temperature and cryogenic freezers.
  • Used with fiberboard cryoboxes to divide box into grids for vial storage
  • Available in 4×4, 5×5, 7×7, 8×8, 9×9 or 10×10 grids