Metre rules
Leverandør: R L ENTERPRISES
Z7904
RLAAZ7904EA 0 NOK
RLAAZ7904
Metre rules
Linjaler
These metre rules are moulded in white plastic 6 mm thick by 25 mm wide and foil printed in black.
  • With metric or with metric and imperial measures