Solvent filter
Leverandør: ROBU GLASFILTER
80052
ROBU80052EA 725 NOK
ROBU80052
Solvent filter
Filter Filter, inntaks-
This solvent filter is made of pure borosilicate glass.
  • Highly resistant to most chemicals such as solvents, acids and alkalis
  • Reduced particle emission, specially rinsed, with calibrated rim