IgG1 Isotype Control (PE (Phycoerythrin))

Leverandør: Rockland Immunochemicals
012-008-040
ROCK012-008-040EA 9500 NOK
ROCK012-008-040
IgG1 Isotype Control (PE (Phycoerythrin))
Antistoffer
Rat IgG1 isotype control can be utilized as a control or standard reagent in Western Blotting, Flow Cytometry, and ELISA experiments where determination of sample isotype is important. Specific conditions should be optimized by user.

Recommended Dilutions: ELISA: 1:2000-1:20000; Flow Cytometry: 1:1000-1:5000;
Do not freeze! Light sensitive!

Type: Control
Antigen: IgG1
Clonality: Monoclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity:
Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Antistofftype Control
Klonalitet Monoclonal
Konjugering PE (Phycoerythrin)
Reaktivitet Drosophila,Plant,Xenopus,Chicken,C. elegans,Human,Rat,Mouse
Vert
Isotype IgG1
ELISA Yes
ImmunoFluorescens Yes
Flowcytometri Yes
Format Antibody fraction
Skjema Liquid (sterile filtered)
Rensing purified by immunoaffinity chromatography
Konsentrasjon 1,0 mg/ml by UV absorbance at 280 nm
Kyssabsorpsjon none
Lagringsbuffer 0,01% (w/v) Sodium Azide,
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR