Anti-CUL5 Rabbit Monoclonal Antibody (Unconjugated) [clone: 14C5]

Leverandør: Rockland Immunochemicals
100-401-A05
ROCK100-401-A05EA 10075 NOK
ROCK100-401-A05
Anti-CUL5 Rabbit Monoclonal Antibody (Unconjugated) [clone: 14C5]
Antistoffer
This antibody reacts with human Cul5 by western blot and immunoprecipitation.  The antibody immunoprecipitates Protein from cell lysates (using 293T, U2OS and others).  This antibody also co-immunoprecipitates associated Proteins.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:1000;

Type: Primary
Antigen: CUL5
Clonality: Monoclonal
Clone: 14C5
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR