Anti-ADH1 Rabbit Polyclonal Antibody

Leverandør: Rockland Immunochemicals
100-4143
ROCK100-4143EA 25250 NOK
ROCK100-4143
Anti-ADH1 Rabbit Polyclonal Antibody
Antistoffer
This antibody has been tested for use in ELISA and by western blot.

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:2000; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: ADH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype:
Reactivity: Yeast
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR