Anti-TNFA Rabbit Polyclonal Antibody

Leverandør: Rockland Immunochemicals
109-401-306
ROCK109-401-306EA 25250 NOK
ROCK109-401-306
Anti-TNFA Rabbit Polyclonal Antibody
Antistoffer
Primary Rabbit Anti-Human TNF alpha Reacts with Human

Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:500 - 1:2000;

Type: Primary
Antigen: TNFA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Primate
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR