Anti-IgG (H+L) Goat Polyclonal Antibody

Leverandør: Rockland Immunochemicals
111-1102
ROCK111-1102EA 3125 NOK
ROCK111-1102
Anti-IgG (H+L) Goat Polyclonal Antibody
Antistoffer
Anti-Rabbit IgG secondary antibody is suitable for use in various immunoassays. Specific conditions for reactivity and signal detection should be optimized by the end user.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR