TrueBlot® IP/Western Blot
Leverandør: Rockland Immunochemicals
13-8817-82 00-8811-25 13-8817-80 88-1488-31 88-1688-31 88-7788-31
ROCK13-8817-82EA 5175 NOK
ROCK13-8817-82 ROCK00-8811-25 ROCK13-8817-80 ROCK88-1488-31 ROCK88-1688-31 ROCK88-7788-31
TrueBlot® IP/Western Blot
Kit til blotting
IP Western blots provide highly specific results, yet often suffer from heavy/light chain blotting, contamination, and ongoing interference.
TrueBlot® reagents enable you to generate clear, best-quality data in your Immunoprecipitation and Western Blot protocols. Available in several options, from IP Beads alone, to complete IP/Western Blot kits from goat, mouse, rabbit or sheep.
TrueBlot® products solve nearly all of these problems through increased sensitivity, less background noise, and enhanced accuracy.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Konsentrasjon 14,4 mg/cc antibody per cc agarose 0,5 mg/ml by UV absorbance at 280 nm
Kyssabsorpsjon No
Reaktivitet arter Mouse Mouse Goat Rabbit Mouse
Frakttemp. Wet Ice Wet Ice Wet Ice Wet Ice Wet Ice
Lagringsbuffer 0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, Proprietary 0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, Proprietary 0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, Proprietary 0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, Proprietary 0,01 M Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, Proprietary
Lagringstemp 4 °C 4 °C